MultiView® (Mouse-HRP/Rabbit-AP) IHC Kit, Enzo Life Sciences
Catalog # 10051-944
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Description:MULTIVIEW® (mouse-HRP/rabbit-AP) IHC kit
- Storage Temperature:+ 4°C
- Processing Time:~2 hours
- Cat. No.:10051-944
- Supplier no.:950-100-0001
- No. of tests:100
Specifications
About this item
Superior multiplex IHC kit for efficient detection of multiple antigens in a single tissue sample.
The MultiView® (mouse-HRP/rabbit-AP) IHC kit is a non-biotin one-step detection system suitable for demonstrating antigens in formalin-fixed paraffin-embedded tissues and cryostat sections. The kit may also be used with blood smears, cytosmears, and cell preparations.
Ordering information: Contains Peroxidase Block, PolyView IHC reagent (mouse-HRP), PolyView IHC reagent (rabbit-AP), HighDef IHC chromogen substrate (DAB, stable) buffer & chromogen, HighDef red IHC chromogen (AP) buffer & chromogen, Empty mixing bottles.
Caution: RUO - Research Use Only