COX Activity Kit, Enzo Life Sciences
Catalog # 89141-666
Supplier: Avantor
CAS Number:
Specifications
- Environmentally Preferable:
- Sensitivity:0.147 Units/mL COX-I (range 7.5 - 60 Units/mL)
- Cat. No.:89141-666
- No. of wells:96 Wells
- Supplier no.:907-003
Specifications
About this item
For the measurement of COX-1 and COX-2 activity.
The COX activity kit is the first commercially available non-radioactive COX activity assay. This kit lets you easily evaluate COX I and COX II inhibitors in a convenient 96-well plate based assay with < 7% intra-assay variation.
Ordering information: Contains Microtiter plate, Chemiluminescent substrate, Ibuprofen, Meloxicam.