ContactUsLinkComponent
IHC Diluent (Primary Ab), Enzo Life Sciences
IHC Diluent (Primary Ab), Enzo Life Sciences
Catalog # 10051-956
Supplier:  Enzo Life Sciences
CAS Number:  
IHC Diluent (Primary Ab), Enzo Life Sciences
Catalog # 10051-956
Supplier:  Enzo Life Sciences
Supplier Number:  950-244-0250
CAS Number:  
Specifications
  • Description:
    IHC Diluent (Primary Ab)
  • Size:
    250 mL
  • Cat. No.:
    10051-956
  • Supplier no.:
    950-244-0250
Specifications
About this item
IHC diluent (primary Ab) can be used as a diluent to prepare primary antibodies and negative controls
Liquid. Primary antibody diluent.
It can be used with any primary antibody irrespective of its source of origin. Primary antibody diluent is specifically designed to stabilize primary antibodies and reduce nonspecific background staining observed during immunostaining.