IHC Tissue Primer, Enzo Life Sciences
10049-364
:
- Description:IHC Tissue Primer
- Size:100 mL
- Cat. No.:10049-364
- Supplier no.:950-234-0100
The IHC tissue primer is a universal enzyme blocking reagent to use in horse radish peroxidase (HRP) and alkaline phosphatase (AP) based immunohistochemical staining procedures done on paraffin tissue sections, frozen tissue sections and cell smears to suppress nonspecific staining due to endogenous enzyme activity.
Liquid. Tissue primer. TRIS-based alkaline solution with 30% H2O2.