15-deoxy-δ12,14-PGJ2 ELISA Kit, Enzo Life Sciences
89141-290
:
- Assay Type:Competitive
- Target protein:15-deoxy-δ12,14-PGJ2
- Sensitivity:36.8 pg/mL (range 195 - 200,000 pg/mL)
- Cat. No.:89141-290
- Supplier no.:901-023
- No. of tests:96 wells
First-to-market 15-d-PGJ2 ELISA commercially available for eicosanoid research.
For the quantitative determination of 15-deoxy-Δ12,14-PGJ2 in culture supernatants, plasma, saliva, and urine from any species.
The 15-deoxy-Δ12,14-PGJ2 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 5 hours. Absorbance is read at 405 nm. This kit saves time and is much less time consuming than GC/MS analysis. Ready-to-use liquid color-coded reagents reduce laboratory error.
: Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-082 (1x96 wells).