IHC Universal Negative Control Reagent, Enzo Life Sciences
Catalog # 10049-764
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Description:IHC Universal Negative Control Reagent
- Size:25 mL
- Cat. No.:10049-764
- Supplier no.:950-231-0025
Specifications
About this item
IHC universal negative control reagent is specifically designed to work with rabbit, mouse and goat antibodies
Liquid. Negative control reagent- mixture of purified goat, mouse and rabbit immunoglobulins (IgGs).
The reagent has been titered to work with polymer-based secondary systems in manual and automated IHC platforms.