ContactUsLinkComponent
Shipper, EHS Ambient, Therapak®
Shipper, EHS Ambient, Therapak®
  10029-394
undefined
Shipper, EHS Ambient, Therapak®
  10029-394
 :  56872
  • Description:
    Small transport box, specimen bag and absorbent
  • Cat. No.:
    10029-394
  • Supplier no.:
    56872
Ambient shipping system is designed for shipping one standard tube or larger specimen containers (30 mL to 60 mL) of liquid specimen
System features: small transport box (ID: 5.25 x 4 x 3.75"; OD: 6 x 4.25 x 4"), specimen transport bag, absorbent material, Exempt Human Specimen label, biohazard symbol label and cushioning material where applicable.