TI-30 Xa Scientific Calculator
Catalog # 470349-970
Supplier: D&H
Specifications
- Description:TI-30 Xa scientific calculator
- Environmentally Preferable:
- Cat. No.:470349-970
Specifications
About this item
This fundamental calculator combines statistics and advanced scientific functions for high school math and science.
- Adds, subtracts, multiplies, and divides fractions entered in traditional numerator/denominator format
- Performs trigonometric functions, logarithms, roots, powers, reciprocals, and factorials
- Polar/rectangular conversions
- One-variable statistics include results for mean and standard deviation
- Battery powered
This scientific calculator is an industry standard, and a workhorse in classrooms around the world. A solid solution for general math, algebra, trigonometry, and statistics.
Ordering information: This calculator includes a slide case.