HIGHDEF® Red IHC Chromogen (HRP), Enzo Life Sciences
10049-360
:
- Description:HighDef® Red IHC Chromogen (HRP)
- Size:30 mL
- Cat. No.:10049-360
- Supplier no.:950-210-0030
The HighDef® IHC chromogen substrate (AEC, stable) can be used in conjunction with peroxidase-based immunostaining systems.
Liquid.
Peroxidase from the immunodetection system reacts with hydrogen peroxide substrate to degrade it, which reacts with AEC, precipitating it at positive sites yielding a reddish brown colored product.