ContactUsLinkComponent
HighDef® IHC Enhancer (DAB, plus), Enzo Life Sciences
HighDef® IHC Enhancer (DAB, plus), Enzo Life Sciences
  10049-346
 :  
HighDef® IHC Enhancer (DAB, plus), Enzo Life Sciences
  10049-346
 :  950-182-0015
 :  
  • Description:
    HighDef® IHC Enhancer (DAB Plus)
  • Size:
    15 mL
  • Cat. No.:
    10049-346
  • Supplier no.:
    950-182-0015
The HighDef® IHC enhancer (DAB, plus) is a DAB signal enhancer that can be used to increase intensity of immunostaining utilizing DAB as chromogen.
Liquid. 1X DAB chromogen enhancer.