Samco™ Bio-Tite™ Specimen Container, 20 ml/35 mm, Thermo Scientific
Catalog # 95043-678
Supplier: Samco
Specifications
- Sterility:Non sterile
- Capacity:20 ml
- Cat. No.:95043-678
- Packaging:Bulk (Cap/Container Bagged Separately)
- Label type:No label
- Cap color:White
- Supplier no.:06 0181
Specifications
About this item
Prevent bio-hazardous spills with the leak-resistant Samco™ Bio-Tite™ 20mL/35mm Specimen Container
- Plug seal cap ensures excellent leak-resiatant performance
- Cap and base knurls ensure ease of handling
- 95kPa compliant to ensure leak-free performance during transportation
This stackable, shatter-resistant container is manufactured from virgin polypropylene, and is ideal for the collection and transportation of both liquid and solid samples. Supplied bulk packed, ideal for pre-fill applications.
Certifications: 95kPa Compliant.
Ordering information: Ambient storage and shipping recommended for containers.