ContactUsLinkComponent
Anti-static Device, MilliporeSigma
Anti-static Device, MilliporeSigma
Catalog # EM1.07070.0001
Supplier:  MilliporeSigma
Anti-static Device, MilliporeSigma
Catalog # EM1.07070.0001
Supplier:  MilliporeSigma
Supplier Number:  1.07070.0001
Specifications
  • Size:
    1 Set
  • Cat. No.:
    EM1.07070.0001
  • Supplier no.:
    1.07070.0001
Specifications
About this item
If flammable liquids (e.g
solvents) are to be withdrawn or filled, the container must be properly grounded according to valid safety regulations to avoid explosions and fire risks. The relevant measures are to be taken to discharge static electricity. This can be done by using the antistatic device set. It contains 3 cables for highest flexibility in grounding containers during all work steps. It comes in a suitcase for safe and professional storage of cables that are currently not in use. Cable length: 2 m.