Shipper, Category B Ambient, 95 kPa Bag, Insulated Envelope, Gel Wrap, Therapak®
Catalog # 10029-322
Supplier: Therapak, LLC
Specifications
- Description:Cat B ambient, 95 kpa bag, insulated envelope, gel wrap
- Cat. No.:10029-322
- Supplier no.:37980
Specifications
About this item
The ambient transport system features a tray-type transport box (ID: 11.25x9x2.125"; OD: 12.25x9.25x2.25"), insulated envelope, gel wrap, 95kPa bag, Aqui-Pak™ segmented absorbent pouch, list of contents card, biohazard symbol label and Biological Substance Category B label.
It holds up to six transport tubes that are placed inside the absorbent pouch and inserted into the 95kPa bag. The bag is then placed in between the gel wrap and inserted into the envelope to guard against temperature spikes during transit. Maintains ambient temperature.