ContactUsLinkComponent
Shipper, Category B Frozen, Medium Canister, Therapak®
Shipper, Category B Frozen, Medium Canister, Therapak®
Catalog # 10029-262
Supplier:  Therapak, LLC
undefined
Shipper, Category B Frozen, Medium Canister, Therapak®
Catalog # 10029-262
Supplier:  Therapak, LLC
Supplier Number:  37806
Specifications
  • Description:
    Cat B frozen, Medium canister
  • Cat. No.:
    10029-262
  • Supplier no.:
    37806
Specifications
About this item
Frozen specimen shipping system includes: transport box (OD: 11x9x11"), small cooler (ID: 8x6x8"), medium 95kPa canister, Aqui-Pak™ segmented absorbent pouch, cushioning material, list of contents card, dry ice label, and required labels for IATA PI650 marking and compliance
Holds up to six tubes for overnight shipments.