Order Entry
United States
ContactUsLinkComponent
Spectroquant® Nitrate Test In Seawater, MilliporeSigma
Spectroquant® Nitrate Test In Seawater, MilliporeSigma
Catalog # EM1.14942.0001
Supplier:  MilliporeSigma
CAS Number:  
Spectroquant® Nitrate Test In Seawater, MilliporeSigma
Catalog # EM1.14942.0001
Supplier:  MilliporeSigma
Supplier Number:  1.14942.0001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Chemical method:
    Resorcinol
  • Method:
    Photometric
  • Storage Temperature:
    Store at +15°C to +25°C.
  • Cat. No.:
    EM1.14942.0001
  • Supplier no.:
    1.14942.0001
  • No. of tests:
    50
  • For unit:
    Prove, Nova 60

Specifications

About this item

This Spectroquant® Nitrate Cell Test allows the accurate quantification of nitrate in seawater samples. Nitrate is an essential nutrient required for the growth of animals and plants. However, elevated levels of nitrate can be a health risk, in particular for pregnant women and infants. Therefore, nitrate levels in water sources are regulated in many countries.

  • Highly stable, ready to use reagent mixtures in a user friendly design
  • Free selection of appropriate cell format according to the measuring range
  • Long shelf life up to 3 years - all at room temperature
  • Automatic selection of the correct method with help of AutoSelector barcode identification

Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.