ContactUsLinkComponent
Advansta AdvanWash™ Washing Solution
Advansta AdvanWash™ Washing Solution
Catalog # 490005-048
Supplier:  Advansta
CAS Number:  
Advansta AdvanWash™ Washing Solution
Catalog # 490005-048
Supplier:  Advansta
Supplier Number:  R-03024-D50
CAS Number:  
Specifications
  • Description:
    AdvanWash
  • Size:
    500 mL
  • Cat. No.:
    490005-048
  • Supplier no.:
    R-03024-D50
Specifications
About this item
AdvanWash is optimized to work with Advansta Western blotting system
Choose AdvanWash for all WesternBright chemiluminescent HRP substrates and the WesternBright MCF fluorescent Western blotting kit.