Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Chemical method:Nitrospectral / sulfuric acid
- Description:Nitrate Test
- Method:Colorimetric with color-disk comparator
- Storage Temperature:Store at +15°C to +25°C.
- Cat. No.:EM1.18387.0001
- Range:5-10-20-30-40-50-60-70-80-90 mg/L NO₃
- Supplier no.:1.18387.0001
- No. of tests:90
Specifications
About this item
Nitrate is naturally present in soil, water and food, and regularly testing its concentration is essential for many applications
When the reagents of the MColortestTM test are used up, the kit can be replenished by ordering the appropriate refill pack. Refill packs are available for a range of MColortestTM test kits, making it a particularly economical system.
Nitrates must be closely monitored in drinking and ground water as well as in process, waste, or aquarium water.