Corning® Gosselin™ Automation Petri Dishes, 100×15 mm, Corning
Catalog # 10416-318
Supplier: Avantor
Specifications
- Material:PS
- Description:Gosselin 100 x 15 mm Automation Petri Dish
- For Use With:Automation
- Packaging:25 per sleeve/500 per case
- Size:100 x 15 mm
- Cat. No.:10416-318
- Supplier no.:BP94A-01
Specifications
About this item
The new Corning® Gosselin™ 100x15 Petri dishes design had been designed in collaboration with Media pourer automation manufacturer.
- Manufactured from optically clear virgin polystyrene
- Sterilized by irradiation SAL 10-3
- Supplied with 4 vents to provide consistent gas exchange
- New design to enhance stacked dishes stability