Rodent Diet, Grain-Based, Control, Bio-Serv
Catalog # 89067-483
Supplier: Bio-Serv
Specifications
- Ø:1.27 cm (½")
- Description:Control Diet
- Height:2.54 cm (1")
- Size:Pellets
- Cat. No.:89067-483
- Packaging:10 kg/Box
- Supplier no.:F3028
Specifications
About this item
Nutritionally complete grain-based diet used as a control when ordering custom grain-based diets
- Nutritionally assayed
- Shelf Life: 6 months, cool dry storage
Ingredients: Ground Corn, Dehulled Soybean Meal, Dried Beet Pulp, Fish Meal, Ground Oats, Brewers Dried Yeast, Cane Molasses, Dehydrated Alfalfa Meal, Dried Whey, Wheat Germ, Porcine Animal Fat (preserved with BHA), Porcine Meat Meal, Wheat Middlings, Mineral Mix, Vitamin Mix, DL-Methionine, Choline Chloride.
Caloric value: 3.35 kcal/g
Nutritional ash content: 7.0%
Nutritional carbohydrate content: 48.7%
Nutritional fat content: 5.0%
Nutritional fibre content: 5.1%
Nutritional moisture content: <10%
Nutritional protein content: 23.9%