Order Entry
United States
ContactUsLinkComponent
Nutra-Gel Diet™ Purified Formula, Rodent, Sterile, Bio-Serv
Nutra-Gel Diet™ Purified Formula, Rodent, Sterile, Bio-Serv
Catalog # 89132-274
Supplier:  Bio-Serv
undefined
Nutra-Gel Diet™ Purified Formula, Rodent, Sterile, Bio-Serv
Catalog # 89132-274
Supplier:  Bio-Serv
Supplier Number:  S5769-TRAY

Specifications

  • Description:
    Nutra-Gel Tray, Bacon Flavor
  • Size:
    40 g/cube
  • Cat. No.:
    89132-274
  • Supplier no.:
    S5769-TRAY

Specifications

About this item

Nutritionally complete, purified, bacon-flavored gel diet that provides both food and water for rodents, and is especially useful in cases involving chewing and mobility impairments, post-op care, weanlings, or transportation

  • Trays contain 16 pre-measured cubes that will pop out as needed and can be placed directly on the bedding or in a food hopper
  • Nutra-Gel remains moist in the cage 24 to 48 hours, depending on the type of cage system
  • Convenient and ready to serve; no mixing required
  • Each cell is individually sealed, which maintains moisture and sterility
  • Phytoestrogen free
  • Bacon flavor stimulates appetite
  • Nutritionally assayed
  • Shelf Life: 6 months frozen, 3 months refrigerated

Ingredients: Water, Sucrose, Casein, Corn Starch, Soybean Oil, Gelatin, Cellulose, Mineral Mix, Vitamin Mix, Bacon Flavor, L-Cystine, Xanthan Gum, Sorbic Acid, Choline Bitartrate, tBHQ.

Caloric value: 1.57 kcal/g
Nutritional ash content: 0.9%
Nutritional carbohydrate content: 22.2%
Nutritional fat content: 3.3%
Nutritional fibre content: 1.9%
Nutritional moisture content: <70%
Nutritional protein content: 9.5%

Ordering information: Most mice consume approximately 10-12 g of Nutra-Gel per day. Rats consume approximately 26-60 g of Nutra-Gel per day, depending on strain and age.