Order Entry
United States
ContactUsLinkComponent
 
Timothy Hay Cubes, Certified, Bio-Serv
  89067-422
 :  Bio-Serv
undefined
Timothy Hay Cubes, Certified, Bio-Serv
  89067-422
 :  Bio-Serv
 :  F1011

 

  • Ø:
    3.18 cm (1¼")
  • Description:
    Timothy Cubes
  • Height:
    1.9 cm (¾")
  • Cat. No.:
    89067-422
  • Packaging:
    8.63 kg/Box
  • Supplier no.:
    F1011

 

 

Contains 100% sun-cured timothy hay, steam pressed

  • No binders added
  • Excellent source of fiber
  • Nutritionally assayed.
  • Shelf Life: 24 months, cool dry storage.

Caloric value: 1.91 kcal/g
Nutritional ash content: 8.5%
Nutritional carbohydrate content: 31.5%
Nutritional fat content: 1.0%
Nutritional fibre content: 35.0%
Nutritional moisture content: <10%
Nutritional protein content: 14.0%

 : Certified (Contaminant Screened).