Order Entry
United States
Orders LinkContactUsLinkComponent
Rabbit Stix™, Certified, Bio-Serv

Rabbit Stix™, Certified, Bio-Serv

Rabbit Stix Pellets, Pineapple, 0.48 cm (³⁄₁₆"), 2.54 cm (1"), 10 kg/Box
Catalog # 89067-420
Supplier:   Bio-Serv
Supplier #:  F2831
Rabbit Stix™, Certified, Bio-Serv
Catalog # 89067-420
Supplier:  Bio-Serv
Rabbit Stix™, Certified, Bio-Serv
Rabbit Stix™, Certified, Bio-Serv

Rabbit Stix Pellets, Pineapple, 0.48 cm (³⁄₁₆"), 2.54 cm (1"), 10 kg/Box

Price
$147.25
Log in for availability
Each (10kg)
Minimum order quantity is 1
Product Details & Documents

High-fiber, pineapple-flavored maintenance diet, supplement or enrichment treat.

  • Contains papain, commonly used to prevent hairballs, and is an excellent source of Vitamin C
  • Use as an appetite stimulant for new arrivals, post-op or for debilitated conditions
  • Also recommended for Guinea Pigs
  • Nutritionally assayed
  • Shelf Life: 6 months, cool dry storage

Ingredients: Alfalfa Meal, Ground Corn, Wheat Middlings, Dehulled Soybean Meal, Ground Oats, Ground Soybean Hulls, Cane Molasses, Soy Protein Isolate, Soybean Oil, Mineral Mix, Vitamin Mix, Choline Chloride, Lubricant, Papain, Ascorbic Acid, Pineapple Flavor, Niacin.

Caloric value: 2.73 kcal/g
Nutritional ash content: 7.6%
Nutritional carbohydrate content: 42.0%
Nutritional fat content: 3.7%
Nutritional fibre content: 22.0%
Nutritional moisture content: <10%
Nutritional protein content: 18.0%

Certifications: Certified (Contaminant Screened).

Specifications
  • Ø:
    0.48 cm (³⁄₁₆")
  • Description:
    Rabbit Stix Pellets, Pineapple
  • Height:
    2.54 cm (1")
  • Packaging:
    10 kg/Box
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.