Order Entry
United States
ContactUsLinkComponent
ReliaPrep™ Non sterile PTFE Syringe Filters, Ahlstrom
ReliaPrep™ Non sterile PTFE Syringe Filters, Ahlstrom
Catalog # 10192-180
Supplier:  Ahlstrom
undefined
ReliaPrep™ Non sterile PTFE Syringe Filters, Ahlstrom
Catalog # 10192-180
Supplier:  Ahlstrom
Supplier Number:  760545

Specifications

  • Ø:
    4 mm
  • Pore size:
    0.45 µm
  • Cat. No.:
    10192-180
  • Supplier no.:
    760545

Specifications

About this item

Ahlstrom offers syringe filters for convenient filtration of small sample sizes through membranes.