Order Entry
United States
ContactUsLinkComponent
Super Challenger™ Enrichment Device, Bio-Serv
Super Challenger™ Enrichment Device, Bio-Serv
Catalog # 89067-782
Supplier:  Bio-Serv
undefined
Super Challenger™ Enrichment Device, Bio-Serv
Catalog # 89067-782
Supplier:  Bio-Serv
Supplier Number:  K3100-S

Specifications

  • Ø:
    16.51 cm (6.5")
  • Description:
    Super Challenger, Yellow
  • Cat. No.:
    89067-782
  • Supplier no.:
    K3100-S

Specifications

About this item

Made of sturdy polyethylene with a 10" stainless steel adjustable chain

Internal disk provides additional challenge. Hole size accommodates larger treats. Hang from outside of cage for easy filling. Cagewasher safe.