Jackson-Pratt® Wound Drains, Cardinal Health
Supplier: Cardinal Health
Certificates
About this item
Jackson-Pratt Flat Drains are made of a silicone elastomer for softness and flexibility.
- Injection molded for strength and drain integrity
- Low-profile, hubless design to help promote tissue plane approximation
- Inner lumen ribbing to help prevent drain collapse and clogging and help preserve drain patency
- Silicone that is barium impregnated for x-ray detection of drain
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.