ContactUsLinkComponent
Universal Chain Assembly for Enrichment Devices, Bio-Serv
Universal Chain Assembly for Enrichment Devices, Bio-Serv
Catalog # 89067-720
Supplier:  Bio-Serv
undefined
Universal Chain Assembly for Enrichment Devices, Bio-Serv
Catalog # 89067-720
Supplier:  Bio-Serv
Supplier Number:  K9005
Specifications
  • Description:
    Universal Chain Assembly
  • Length:
    34.9 cm (13.75")
  • Cat. No.:
    89067-720
  • Supplier no.:
    K9005
Specifications
About this item
A convenient way to suspend enrichment products
Made of stainless steel. Cagewasher safe.