RDE II Haemagglutination Test for Influenza
Catalog # 10753-482
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Vial
- Size:20 ml
- Enzyme Name:RDE II
- Cat. No.:10753-482
- Supplier no.:370013
Specifications
About this item
Used for eliminating inhibitors in human serum during hemagglutination inhibition testing of the influenza virus.
Ordering information: RDE II is for research use only.
Packaging: 5 vials