HardyDisk™ AST Azithromcin, AZM-15, Hardy Diagnostics
Catalog # 10753-480
Supplier: Hardy Diagnostics
Product available on GSA Advantage®
CAS Number:
Specifications
- Description:HardyDisk Azithromycin, AZM-15
- Cat. No.:10753-480
- Supplier no.:Z8051
Specifications
About this item
HardyDisk™ ASTs are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST), also known as disk diffusion or Kirby-Bauer testing.
Ordering information: Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.