Order Entry
United States
ContactUsLinkComponent
Somso® Eyeball with Part of Orbit
Somso® Eyeball with Part of Orbit
  470159-602
undefined
Somso® Eyeball with Part of Orbit
  470159-602
 :  CS 2

 

  • Description:
    Eyeball with part of orbit
  • Height:
    23 cm
  • Length:
    19 cm
  • Cat. No.:
    470159-602
  • Width:
    32 cm
  • Supplier no.:
    CS 2

 

 

Somso® premium quality models.

  • Five year warranty

Enlarged approx. three times, in SOMSO-Plast®. All six muscles of the eye are represented. The optic nerve is shown up to where it enters the green base of the skull, the eyeball is sectioned horizontally. Separates into eight parts: superior rectus and exterior straight muscle, upper half of the sclerotic membrane, choroid membrane and retina (2), vitreous humour, lens, orbit with green base.

 : Items on this page are not stocked and may have an extended time for delivery.