AlphaTec® 88-393 Medium-Duty Natural Rubber Latex Gloves, Unsupported, Unlined, Ansell
Supplier: Avantor
Certificates
About this item
Type 88-393 unsupported, unlined, medium-duty gloves feature excellent dexterity, tensile strength, and elasticity.
- Hand-specific
- Provide good chemical, snag, puncture, abrasion, and cut resistance
Gloves are suitable for poultry, food processing, small parts assembly, pharmaceutical manufacturing, and janitorial/plant maintenance.
Certifications: Gloves are made with component materials that comply with FDA regulations for food contact.
Packaging: Packaged 12 pairs in a bag; 144 in a carton.
Caution: Gloves contain natural rubber latex, which may cause allergic reactions in some individuals.
Specifications
- Catalog No:
- 80092-568
- 80092-562
- 80092-566
- 80092-564
- Ambidextrous/Hand-specific:
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Finish:
- Fishscale
- Fishscale
- Fishscale
- Fishscale
- Cuff Style:
- Scalloped
- Scalloped
- Scalloped
- Scalloped