ContactUsLinkComponent
ActivArmr® 80-813 Foam Neoprene Flame Resistant Gloves, Ansell
ActivArmr® 80-813 Foam Neoprene Flame Resistant Gloves, Ansell
Supplier:  Ansell Healthcare
Certificates
undefined
ActivArmr® 80-813 Foam Neoprene Flame Resistant Gloves, Ansell
Supplier:  Ansell Healthcare
Specifications
  • Catalog No:
  • 89413-052
  • 89413-042
  • 89413-048
  • 89413-050
  • 89413-044
  • 89413-046
  • Ambidextrous/Hand-specific:
  • Hand-Specific
  • Hand-Specific
  • Hand-Specific
  • Hand-Specific
  • Hand-Specific
  • Hand-Specific
  • Liner Material:
  • Modacrylic/glass fibre/Kevlar®
  • Modacrylic/glass fibre/Kevlar®
  • Modacrylic/glass fibre/Kevlar®
  • Modacrylic/glass fibre/Kevlar®
  • Modacrylic/glass fibre/Kevlar®
  • Modacrylic/glass fibre/Kevlar®
  • Finish:
  • Palm Coated
  • Palm Coated
  • Palm Coated
  • Palm Coated
  • Palm Coated
  • Palm Coated
  • Cuff Style:
  • Knit Wrist
  • Knit Wrist
  • Knit Wrist
  • Knit Wrist
  • Knit Wrist
  • Knit Wrist
  • EN Norm:
  • 2542
  • 2542
  • 2542
  • 2542
  • 2542
  • 2542