ContactUsLinkComponent
Hepatitis C (HCV) PBMC, Charles River Laboratories Cell Solutions
Hepatitis C (HCV) PBMC, Charles River Laboratories Cell Solutions
Catalog # 102830-116
Hepatitis C (HCV) PBMC, Charles River Laboratories Cell Solutions
Catalog # 102830-116
Supplier Number:  PB009C-1-HCV
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Format:
    Cryopreserved
  • Description:
    Hepatitis C (HCV) Mononuclear Cells (PBMC)
  • Size:
    10 x 10e6 Cells
  • Cat. No.:
    102830-116
  • Supplier no.:
    PB009C-1-HCV
Specifications
About this item
Human disease state peripheral blood mononuclear cells (PBMC) are collected from human volunteer donors under IRB approved informed consent by whole blood or leukapheresis using the COBE® Spectra Apheresis System MNC collection protocol
Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.
This mononuclear cell-rich leukapheresis product (LeukoPak) is extracted using continuous flow centrifugal technology directly into the collection bag. Mononuclear cells are purified using density gradient centrifugation techniques.