Specifications
- Format:Cryopreserved
- Description:Type 2 Diabetes Mononuclear Cells (PBMC)
- Size:10 x 10e6 Cells
- Cat. No.:102830-178
- Supplier no.:PB009C-1-DM2
Specifications
About this item
Human disease state peripheral blood mononuclear cells (PBMC) are collected from human volunteer donors under IRB approved informed consent by whole blood or leukapheresis using the COBE® Spectra Apheresis System MNC collection protocol
Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.
This mononuclear cell-rich leukapheresis product (LeukoPak) is extracted using continuous flow centrifugal technology directly into the collection bag. Mononuclear cells are purified using density gradient centrifugation techniques.