ContactUsLinkComponent
Lung Cancer Serum, Charles River Laboratories Cell Solutions
Lung Cancer Serum, Charles River Laboratories Cell Solutions
Catalog # 102830-136
Lung Cancer Serum, Charles River Laboratories Cell Solutions
Catalog # 102830-136
Supplier Number:  PB007C-1-LC
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Serum format:
    Cryopreserved
  • Description:
    Lung Cancer Serum
  • Size:
    500 mL
  • Environmentally Preferable:
  • Cat. No.:
    102830-136
  • Supplier no.:
    PB007C-1-LC
Specifications
About this item
Human disease state peripheral blood serum is collected in HemaCare's FDA-registered collection centers or through our hospital or physician office network from IRB consented human volunteer donors
Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.
Serum is collected from peripheral blood in a plain or Serum Separator Tube.