Order Entry
United States
ContactUsLinkComponent
Gastric/Stomach Cancer Plasma, Charles River Laboratories Cell Solutions
Gastric/Stomach Cancer Plasma, Charles River Laboratories Cell Solutions
Catalog # 102830-182
CAS Number:  
Gastric/Stomach Cancer Plasma, Charles River Laboratories Cell Solutions
Catalog # 102830-182
Supplier Number:  PB006C-1-GC
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Gastric/Stomach Cancer Plasma
  • Size:
    2 mL
  • Cat. No.:
    102830-182
  • Format:
    Cryopreserved
  • Supplier no.:
    PB006C-1-GC

Specifications

About this item

Human disease state peripheral blood plasma is collected in HemaCare's FDA-registered collection centers or through our hospital or physician office network from IRB consented human volunteer donors

Serological and Infectious disease testing are performed with each donation. Customers should handle product as if potentially infectious following biosafety level 2 procedures.

By default, plasma is separated from whole blood which has been collected in a CPD collection bag or EDTA specimen tubes. If possible, HemaCare will utilize any commercially available special collection tubes with anticoagulants per your specifications.