ContactUsLinkComponent
Giemsa's stain solution, Sigma-Aldrich®
Flammable
Toxic
Giemsa's stain solution, Sigma-Aldrich®
Supplier:  MilliporeSigma
Certificates
Flammable
Toxic
Giemsa's stain solution, Sigma-Aldrich®
Supplier:  MilliporeSigma
Certificates
Specification Test Results
  • Chemical formula:
    Mixture
  • Key Applications:
    General Chemistry
  • Absorbance at maximum (Azure):
    0.64-0.77AU
  • Absorbance maximum wavelength (Azure):
    648-652nm
  • Absorbance maximum wavelength (Eosin):
    525-528nm
  • Performance:
    Passes
  • Shape of Scan:
    Conforms