Order Entry
United States
ContactUsLinkComponent
Phenol liquified ≥90% in water, Sigma-Aldrich®
Toxic
Corrosive
Health
Phenol liquified ≥90% in water, Sigma-Aldrich®
Catalog # EM8.43984.1000
Supplier:  MilliporeSigma
CAS Number:  
Toxic
Corrosive
Health
Phenol liquified ≥90% in water, Sigma-Aldrich®
Catalog # EM8.43984.1000
Supplier:  MilliporeSigma
Supplier Number:  8.43984.1000
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    EM8.43984.1000
  • Supplier no.:
    8.43984.1000

Specifications

Specification Test Results

  • Density:
    1.07 g/cm³ (25 °C)
  • Flash point:
    81 °C Information refers to the main component.
  • pH value:
    (H₂O) acidic
  • WGK:
    WGK 2 water endangering
  • R Phrase:
    R 23/24/25-34-68-48/20/21/22
  • S Phrase:
    S 24/25-26-28-36/37/39-45

Specification Test Results

About this item

 :
 :94.11 g/mol
ADR:6.1,II
 :M-Clarity™ quality level = MQ200
UN:2821
 :GHS06
 :GHS05
 :GHS08
CAS Number:108-95-2
Formula:C₆H₅OH
MDL Number:MFCD00002143