Formic acid 0,1% in water mobile phase for HPLC, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
CAS Number:64-18-6
:
Formula:HCO₂H
MDL Number:MFCD00003297
:46.03
UN:1779
ADR:8,II
Specification Test Results
- Expiration date:12 months for date of maufacture
- Assay (HCOOH):0.095-0.105% v/v
- Density at 20°C:approximately 0.9985g/cm³
- Form:Clear liquid
- Gradient at 254 nm:0.005AU max
- Gradient at 280 nm:0.003AU max
- UV Absorbance at 220 nm:1.0AU max
- UV Absorbance at 230 nm:0.5AU max
- UV Absorbance at 240 nm:0.10AU max
- UV Absorbance at 250 nm:0.02AU max
- UV Absorbance at 280 nm:0.01AU max
Specification Test Results
Related Information
Frequently Bought Together