Remove-iT PNGase F
Supplier: New England Biolabs (NEB)
Certificates
About this item
Remove-iT® PNGase F is an amidase which cleaves between the innermost GlcNAc and asparagine residues of high mannose, hybrid, and complex oligosaccharides from N-linked glycoproteins
Remove-iT PNGase F is tagged with a chitin binding domain (CBD) for easy removal from a reaction and is supplied glycerol free for optimal performance in HPLC and MS intensive methods.
Specifications
- Enzyme Name:Remove-iT PNGase F