Absorbance is read in the blue spectral range (650 nm).
- 3 year shelf life
A dry blend stop reagent used in end point ELISA applications where a
TMB substrate is used as the detection mechanism.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Conjugate ELISA:HRP
- Color Development:Blue Solute
- Shelf Life:3 years