Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Chemiluminescent Ultra Sensitive AP Microwell and Membrane Substrate (450 nm)
- Size:100 mL
- Shelf Life:2 years
- Storage Temperature:2 to 8 °C
- Application:ELISA, Western Blot, Other Membrane
Applications - Cat. No.:101396-882
- Supplier no.:APU4-0100-01
Specifications
About this item
Chemiluminescent substrates provide a higher degree of sensitivity than other substrates to enable lower limits of detection to be achieved in ELISA and membrane assays. The increase in sensitivity requires more vigilance during the optimization process through the proper selection of the dioxetane substrate, antibody/antigen titers and blockers. Surmodics provides multiple 1-component AP chemiluminescent substrates with various intensities and wavelength emissions to allow the assay developer to choose the appropropriate substrate for the application.
- Absorbance at 450 nm
- High signal to noise ratios
A one-component dioxetane chemiluminescent substrate that provides ultra-sensitive detection of alkaline phosphatase in ELISA and membrane applications.