Order Entry
United States
ContactUsLinkComponent
BioFX® Chemiluminescent Super Sensitive Plus HRP Substrate, Surmodics
BioFX® Chemiluminescent Super Sensitive Plus HRP Substrate, Surmodics
Catalog # 101396-972
Supplier:  Surmodics
CAS Number:  
BioFX® Chemiluminescent Super Sensitive Plus HRP Substrate, Surmodics
Catalog # 101396-972
Supplier:  Surmodics
Supplier Number:  LUMI-0110-2C
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Chemiluminescent Super Sensitive HRP Microwell/Membrane
  • Size:
    110 ml
  • Enzyme Name:
    Horseradish Peroxidase
  • Shelf Life:
    1.5 years
  • Storage Temperature:
    2...8 °C
  • Application:
    ELISA, Western Blot and Other Membrane Substrates
  • Cat. No.:
    101396-972
  • Supplier no.:
    LUMI-0110-2C

Specifications

About this item

BioFX® Chemiluminescent Super Sensitive Plus HRP Substrate provides a higher degree of sensitivity in the detection of peroxidase than other substrates to enable lower limits of detection to be achieved in ELISA and Western Blot assays.

  • Picogram to femtogram levels of sensitivity
  • High signal to noise ratios

The increase in sensitivity requires more vigilance during the optimization process through the proper selection of the luminol substrate, antibody/antigen titers and blockers.

Surmodics provides multiple 2-component HRP chemiluminescent substrates with various intensities to allow the assay developer to choose the appropropriate substrate for the application.

Certifications: ISO 13485:2016 and 9001:2015 certified.