Order Entry
United States
ContactUsLinkComponent
BioFX® Chemiluminescent Ultra Sensitive HRP Microwell Substrate, Surmodics IVD, Inc.
BioFX® Chemiluminescent Ultra Sensitive HRP Microwell Substrate, Surmodics IVD, Inc.
Catalog # 100359-144
Supplier:  Surmodics
CAS Number:  
BioFX® Chemiluminescent Ultra Sensitive HRP Microwell Substrate, Surmodics IVD, Inc.
Catalog # 100359-144
Supplier:  Surmodics
Supplier Number:  CHMI-0060-2C
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Chemiluminescent Ultra Sensitive HRP Microwell Substrate
  • Size:
    60 ml
  • Shelf Life:
    1.5 years
  • Application:
    Microwell
  • Cat. No.:
    100359-144
  • Supplier no.:
    CHMI-0060-2C

Specifications

About this item

Chemiluminescent substrates provide a higher degree of sensitivity than other substrates to enable lower limits of detection to be achieved in Western Blot assays. The increase in sensitivity requires more vigilance during the optimization process through the proper selection of the luminol substrate, antibody/antigen titers and blockers.

  • Higher degree of sensitivity
  • Lower limits of detection

Surmodics provides multiple 2-component HRP chemiluminescent substrates with various intensities to allow the assay developer to choose the appropropriate substrate for the application.

Chemiluminescent substrates enables lower limits of detection to be achieved in ELISA assays. The increase in sensitivity requires more vigilance during the optimization process through the proper selection of the luminol substrate, antibody/antigen titers and blockers.

Certifications: ISO 13485:2016 and 9001:2015 certified.