Order Entry
United States
ContactUsLinkComponent
BioFX® BCIP/NBT Purple One Component AP Membrane Substrate, Surmodics IVD, Inc.
BioFX® BCIP/NBT Purple One Component AP Membrane Substrate, Surmodics IVD, Inc.
Supplier:  Surmodics
Certificates
BioFX® BCIP/NBT Purple One Component AP Membrane Substrate, Surmodics IVD, Inc.
Supplier:  Surmodics

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Conjugate ELISA:
    Alkaline Phosphatase (AP)
  • Storage Conditions:
    2...25 °C
  • Color Development:
    Purple precipitate
  • Shelf Life:
    3 years
  • Application:
    Western Blot and Other Membrane Applications

Specifications

Frequently Bought Together