About this item
The ABTS (2,2’-azino-bis [3-ethylbenzthiazoline-6-sulfonic acid]) substrate is supplied as a one component, ready-to-use solution for ELISA applications. A soluble blue-green reaction product is obtained when this substrate system is reacted with horseradish peroxidase.
- 3 year shelf life
- 1-Component format
- Ready to use formulation
ABTS One Component HRP Microwell Substrate offers sensitive detection of
horseradish peroxidase in microwell applications.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Host:
- Primary antibody reactivity:
- Environmentally Preferable: