To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Description:DAB Immunohistochemistry Substrate
- Size:100 ml
- Color Development:Brown precipitate
- Shelf Life:3 years
- Storage Temperature:2...8 °C
- Application:Immunohistochemistry (IHC)
- Cat. No.:100359-228
- Supplier no.:DABI-0100-2C
DAB (3,3’-diaminobenzidine) is oxidized in the presence of peroxidase and hydrogen peroxide resulting in the deposition of a brown, alcohol-insoluble precipitate at the site of enzymatic activity.
- Three year shelf life
- Rapid and simple detection of peroxidase in immunohistochemical and in situ hybridization procedures
BioFX® DAB Immunohistochemistry Substrate is a two-component substrate system.