Order Entry
United States
ContactUsLinkComponent
Anti-RGSL1 Rabbit Polyclonal Antibody
Anti-RGSL1 Rabbit Polyclonal Antibody
Catalog # 10155-786
Supplier:  Proteintech
undefined
Anti-RGSL1 Rabbit Polyclonal Antibody
Catalog # 10155-786
Supplier:  Proteintech
Supplier Number:  25836-1-AP-150UL

Specifications

  • Antibody Type:
    Primary
  • Antigen Symbol:
    RGSL1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Storage Buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Storage Temperature:
    Store at –20 °C
  • Immunogen:
    RGSL1
  • Purification:
    Antigen affinity purification
  • Cat. No.:
    10155-786
  • Supplier no.:
    25836-1-AP

Specifications

About this item

The RGSL1 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human RGSL1. This antibody recognizes human, mouse antigen. The RGSL1 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 600

Type: Primary
Antigen: RGSL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse