Anti-CLDN23 Rabbit Polyclonal Antibody
Catalog # 10156-132
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Claudin 23
- Antigen Symbol:CLDN23
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage Temperature:Store at –20 °C
- Immunogen:Claudin 23
- Purification:Antigen affinity purification
- Cat. No.:10156-132
- Supplier no.:25296-1-AP
Specifications
About this item
The Claudin 23 Antibody from Proteintech is a rabbit polyclonal Antibody to a fusion protein of human Claudin 23. This Antibody recognizes human Antigen. The Claudin 23 Antibody has been validated for the following applications: ELISA, IHC, WB analysis.
WB 1 : 600
Type: Primary
Antigen: CLDN23
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: