Anti-SNX32 Rabbit Polyclonal Antibody
Catalog # 10155-774
Supplier: Proteintech
Specifications
- Antibody Type:Primary
- Antigen Name:Sorting Nexin 32
- Antigen Symbol:SNX32
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:150 µL
- Antigen Synonyms:SNX6B
- Storage Buffer:PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
- Storage Temperature:Store at –20 °C
- Immunogen:SNX32
- Purification:Antigen affinity purification
- Cat. No.:10155-774
- Supplier no.:25763-1-AP
Specifications
About this item
The SNX32 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human SNX32. This antibody recognizes human antigen. The SNX32 antibody has been validated for the following applications: ELISA, WB analysis.
IHC 1 : 50
Type: Primary
Antigen: SNX32
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human