Order Entry
United States
ContactUsLinkComponent
Anti-MARVELD3 Rabbit Polyclonal Antibody
Anti-MARVELD3 Rabbit Polyclonal Antibody
Catalog # 10155-746
Supplier:  Proteintech
undefined
Anti-MARVELD3 Rabbit Polyclonal Antibody
Catalog # 10155-746
Supplier:  Proteintech
Supplier Number:  25667-1-AP-150UL

Specifications

  • Antibody Type:
    Primary
  • Antigen Name:
    MARVEL Domain Containing 3
  • Antigen Symbol:
    MARVELD3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    150 µL
  • Antigen Synonyms:
    MRVLDC3|MARVD3
  • Storage Buffer:
    PBS with 0.1% sodium azide and 50% glycerol pH 7.3.
  • Storage Temperature:
    Store at –20 °C
  • Immunogen:
    MARVELD3
  • Purification:
    Antigen affinity purification
  • Cat. No.:
    10155-746
  • Supplier no.:
    25667-1-AP

Specifications

About this item

The MARVELD3 antibody from Proteintech is a rabbit polyclonal antibody to a fusion protein of human MARVELD3. This antibody recognizes human antigen. The MARVELD3 antibody has been validated for the following applications: ELISA, WB analysis.

WB 1 : 300 ; IP 1 : 300

Type: Primary
Antigen: MARVELD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human